Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dip2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081426.2 | NP_001074895.2 | 26, 27 | 3689 | ||
GenBank mRNA | AK036188.1 | - | 5, 6 | 544 | ||
AK173066.1 | - | 18, 19 | 2101 | |||
BC056940.1 | - | 9, 10 | 1153 | |||
BC066797.1 | - | 6, 7 | 735 | |||
BC089303.1 | - | 6, 7 | 713 | |||
BC137787.1 | - | 26, 27 | 3281 | 8860806 | ||
BC144909.1 | - | 25, 26 | 3194 | 9053457 | ||
BC144910.1 | - | 26, 27 | 3281 | 9053458 |