Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198019.2 | NP_932136.2 | 8 | 1109 | ||
GenBank mRNA | AK050870.1 | - | 7 | 1235 | ||
AK160295.1 | - | 1 | 340 | |||
BC070400.1 | - | 8 | 1109 | 5694341 | ||
BC071270.1 | - | 8 | 1091 |