Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153782.1 | NP_722477.1 | 10 | 1855 | ||
GenBank mRNA | AK142932.1 | - | 10 | 1880 | ||
BC011274.1 | - | 10 | 1759 | |||
BC023310.1 | - | 3 | 156 | |||
BC029169.1 | - | 10 | 1855 | 4947019 |