Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277992.1 | NP_001264921.1 | 7 | 987 | ||
NM_011494.5 | NP_035624.3 | 7 | 1015 | |||
GenBank mRNA | AB017197.1 | - | 7 | 963 | ||
AB022179.1 | - | 7 | 945 | |||
AF062076.1 | - | 6 | 702 | |||
AF089869.1 | - | 7 | 992 | |||
AJ005790.1 | - | 7 | 946 | |||
AK030404.1 | - | 7 | 990 | |||
AK032162.1 | - | 7 | 1006 | |||
AK149539.1 | - | 7 | 988 | |||
AK156554.1 | - | 6 | 1070 | |||
AK177618.1 | - | 1 | 60 | |||
BC005703.1 | - | 7 | 994 | 3598470 | ||
BC028999.1 | - | 7 | 974 | 5250877 |