Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133670.1 | NP_598431.1 | 4, 5 | 426 | ||
GenBank mRNA | BC005413.1 | - | 4, 5 | 426 | 3488459 | |
L02331.1 | - | 4, 5 | 442 |