Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011499.3 | NP_035629.2 | 6, 7 | 914 | ||
GenBank mRNA | AF096285.1 | - | 7, 8 | 726 | ||
AK075804.1 | - | 6, 7 | 872 | |||
AK077504.1 | - | 6, 7 | 866 | |||
AK145971.1 | - | 6, 7 | 866 | |||
AK147179.1 | - | 6, 7 | 916 | |||
AK152283.1 | - | 6, 7 | 764 | |||
AK154158.1 | - | 6, 7 | 882 | |||
AK210373.1 | - | 2, 3 | 160 | |||
BC141076.1 | - | 6, 7 | 865 | 9055688 | ||
BC145342.1 | - | 6, 7 | 865 | 9053898 |