Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271724.1 | NP_001258653.2 | 7 | 882 | ||
NM_013677.2 | NP_038705.2 | 7 | 918 | |||
GenBank mRNA | AK002469.1 | - | 7 | 732 | ||
AK154726.1 | - | 7 | 738 | |||
AK208539.1 | - | 3 | 261 | |||
BC004755.1 | - | 7 | 918 | 3584373 | ||
BC052500.1 | - | 7 | 905 | 5255619 | ||
CB524655.1 | - | 2 | 188 |