Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201638.2 | NP_964000.2 | 10 | 1078 | ||
GenBank mRNA | AK089949.1 | - | 10 | 1025 | ||
AK146881.1 | - | 10 | 1067 | |||
AK153810.1 | - | 10 | 1074 | |||
BC044899.1 | - | 1 | 1835 | |||
BC052204.1 | - | 10 | 1036 | 30007932 |