Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081396.2 | NP_001074865.2 | 1 | 145 | ||
NM_001167679.1 | NP_001161151.1 | 1 | 145 | |||
GenBank mRNA | AB185844.1 | - | 1 | 113 | ||
AK154264.1 | - | 1 | 110 | |||
AK171024.1 | - | 1 | 110 | |||
BC050039.1 | - | 1 | 91 | 30016266 | ||
BC066830.1 | - | 1 | 77 | 30463017 |