Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253731.1 | NP_001240660.1 | 9, 10 | 1205 | ||
NM_153414.4 | NP_700463.2 | 9, 10 | 1173 | |||
GenBank mRNA | AK038979.1 | - | 9, 10 | 906 | ||
AK077634.1 | - | 9, 10 | 1174 | |||
AK136992.1 | - | 9, 10 | 900 | |||
AK150195.1 | - | 9, 10 | 900 | |||
BC028953.1 | - | 8, 9 | 863 | 5362016 | ||
BC055700.1 | - | 9, 10 | 872 | 6808816 |