Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145960.4 | NP_666072.3 | 3 | 737 | ||
GenBank mRNA | AK043981.1 | - | 3 | 543 | ||
AK051834.1 | - | 3 | 523 | |||
AK077749.1 | - | 3 | 738 | |||
BC028898.1 | - | 3 | 653 | 4205917 |