Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc64b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153784.2 | NP_722479.1 | 6 | 900 | ||
GenBank mRNA | AK036793.1 | - | 6 | 915 | ||
AK133729.1 | - | 6 | 917 | |||
BC038613.1 | - | 6 | 926 | 4458084 |