Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009311.2 | NP_033337.1 | 6 | 411 | ||
GenBank mRNA | BC117081.1 | - | 6 | 386 | 40126400 | |
BC119122.1 | - | 6 | 386 | 8733871 | ||
BC144737.1 | - | 5 | 341 | 9053285 | ||
D17584.1 | - | 6 | 411 | |||
M68908.1 | - | 5 | 207 | |||
M68909.1 | - | 4 | 162 |