Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153136.4 | NP_694776.2 | 3 | 631 | ||
GenBank mRNA | AK089446.1 | - | 2 | 419 | ||
AK137014.1 | - | 3 | 845 | |||
AK145997.1 | - | 3 | 620 | |||
AK155086.1 | - | 3 | 632 | |||
AK157937.1 | - | 3 | 632 | |||
BC036718.1 | - | 3 | 787 | 5322150 |