Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172518.3 | NP_766106.2 | 2 | 507 | ||
GenBank mRNA | AK020135.1 | - | 1 | 418 | ||
AK028867.1 | - | 2 | 353 | |||
AK150804.1 | - | 2 | 498 | |||
AK151503.1 | - | 2 | 464 | |||
AK165284.1 | - | 2 | 481 | |||
BC058667.1 | - | 2 | 498 | 6410012 |