Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081230.1 | NP_001074699.1 | 11 | 1711 | ||
GenBank mRNA | AK135702.1 | - | 8 | 1205 | ||
AK135823.1 | - | 11 | 1615 | |||
AK138540.1 | - | 11 | 1656 | |||
AK147287.1 | - | 11 | 1654 | |||
AK147416.1 | - | 11 | 1654 | |||
AK162555.1 | - | 10 | 1541 | |||
BC051460.1 | - | 3 | 236 | |||
BC061216.1 | - | 11 | 1567 | |||
BC087935.1 | - | 11 | 1608 |