Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081230.1 | NP_001074699.1 | 3 | 378 | ||
GenBank mRNA | AK030527.1 | - | 3 | 219 | ||
AK038737.1 | - | 3 | 323 | |||
AK135823.1 | - | 3 | 282 | |||
AK138540.1 | - | 3 | 323 | |||
AK147287.1 | - | 3 | 322 | |||
AK147416.1 | - | 3 | 322 | |||
AK162555.1 | - | 3 | 298 | |||
BC061216.1 | - | 3 | 234 | |||
BC087935.1 | - | 3 | 275 |