Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145394.3 | NP_663369.2 | 10 | 1374 | ||
GenBank mRNA | BC010552.1 | - | 9 | 1142 | 3582543 | |
BC025548.1 | - | 10 | 1372 | 5320841 |