Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033253.3 | NP_001028425.1 | 10 | 1432 | ||
NM_001159942.1 | NP_001153414.1 | 11 | 1598 | |||
GenBank mRNA | AK122468.1 | - | 7 | 756 | ||
AK135923.1 | - | 10 | 1434 | |||
BC076628.1 | - | 8 | 829 | |||
BC117965.1 | - | 9 | 1034 | 40098326 | ||
BC117966.1 | - | 9 | 1027 | 40098330 |