Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177460.4 | NP_803411.3 | 6 | 1048 | ||
GenBank mRNA | AK033791.1 | - | 998 | |||
AK133475.1 | - | 6 | 1049 | |||
AK166002.1 | - | 5 | 813 | |||
AK169866.1 | - | 7 | 1068 | |||
BC002197.1 | - | 2 | 191 | |||
BC014731.1 | - | 5 | 1078 | |||
BC055447.1 | - | 6 | 1017 | 3594406 |