Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153541.3 | NP_705769.2 | 2 | 1019 | ||
GenBank mRNA | AK132152.1 | - | 2 | 1057 | ||
AK140914.1 | - | 2 | 1008 | |||
AK163930.1 | - | 2 | 1020 | |||
AK163989.1 | - | 2 | 1023 | |||
BC023839.1 | - | 2 | 1014 | 5344921 |