Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172546.2 | NP_766134.1 | 1 | 589 | ||
GenBank mRNA | AK033015.1 | - | 1 | 590 | ||
AY151137.1 | - | 1 | 551 |