Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfc3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033261.2 | NP_001028433.2 | 33 | 5990 | ||
GenBank mRNA | AK133182.1 | - | 5 | 581 | ||
AK161448.1 | - | 5 | 559 | |||
AK172969.1 | - | 31 | 4690 | |||
BC033596.1 | - | 11 | 1320 | |||
BC046488.1 | - | 19 | 2835 | |||
BC061510.1 | - | 21 | 3243 | |||
BC139124.1 | - | 33 | 5847 | 8862146 |