Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011562.2 | NP_035692.2 | 5 | 688 | ||
GenBank mRNA | AK133730.1 | - | 5 | 626 | ||
AK163969.1 | - | 6 | 687 | |||
BC052646.1 | - | 5 | 630 | 30029798 | ||
M87321.1 | - | 5 | 597 |