Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166669.1 | NP_001160141.1 | 19 | 2747 | ||
NM_001166670.1 | NP_001160142.1 | 19 | 2744 | |||
NM_001166671.1 | NP_001160143.1 | 19 | 2626 | |||
NM_172558.3 | NP_766146.2 | 19 | 2747 | |||
GenBank mRNA | AK049216.1 | - | 19 | 2682 | ||
AK131137.1 | - | 19 | 2599 | |||
AK143536.1 | - | 19 | 2696 | |||
AK162955.1 | - | 6 | 715 | |||
AK164214.1 | - | 7 | 822 | |||
BC137802.1 | - | 19 | 2690 | 8860821 | ||
BC144921.1 | - | 19 | 2682 | |||
BC144922.1 | - | 19 | 2690 | 9053470 |