Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017426.1 | NP_001017426.1 | 19 | 4737 | ||
GenBank mRNA | AK129121.1 | - | 9 | 1529 | ||
AK136019.1 | - | 9 | 918 | |||
BC038313.1 | - | 4 | 375 | |||
BC075632.1 | - | 20 | 4913 | 30362899 |