Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017426.1 | NP_001017426.1 | 21 | 5082 | ||
GenBank mRNA | AK129121.1 | - | 11 | 1874 | ||
AK136019.1 | - | 11 | 1263 | |||
BC029201.1 | - | 2 | 176 | |||
BC038313.1 | - | 6 | 720 | |||
BC075632.1 | - | 22 | 5258 | 30362899 |