Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153788.3 | NP_722483.2 | 19 | 2137 | ||
GenBank mRNA | AK133473.1 | - | 14 | 3335 | ||
AK138736.1 | - | 19 | 2130 | |||
AK154449.1 | - | 19 | 2140 | |||
AK155001.1 | - | 19 | 2158 | |||
AK220213.1 | - | 2 | 191 | |||
BC031462.1 | - | 19 | 2137 | 4017986 | ||
BC037481.1 | - | 14 | 1764 | 4460639 | ||
BC064116.1 | - | 5 | 1420 |