Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282006.1 | NP_001268935.1 | 6, 7 | 1012 | ||
NM_001282007.1 | NP_001268936.1 | 6, 7 | 961 | |||
NM_011569.3 | NP_035699.2 | 6, 7 | 964 | |||
GenBank mRNA | AF081947.1 | - | 6, 7 | 955 | ||
AK005801.1 | - | 6, 7 | 1013 | |||
BC050967.1 | - | 6, 7 | 962 | |||
BC098181.1 | - | 6, 7 | 922 | 6312556 |