Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167680.1 | NP_001161152.1 | 12, 13 | 1782 | ||
NM_172572.3 | NP_766160.2 | 12, 13 | 1996 | |||
GenBank mRNA | AK028872.1 | - | 12, 13 | 1997 | ||
AK154639.1 | - | 12, 13 | 1762 | |||
AK171085.1 | - | 12, 13 | 1778 | |||
AY167043.1 | - | 12, 13 | 1782 | |||
BC031398.1 | - | 6, 7 | 815 | |||
BC052182.1 | - | 12, 13 | 1730 | 6335022 |