Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009354.1 | NP_033380.1 | 7 | 2320 | ||
GenBank mRNA | AF051911.1 | - | 7 | 2320 | ||
AF073311.1 | - | 7 | 2291 | |||
BC082327.1 | - | 8 | 1822 | |||
BC127068.1 | - | 8 | 1608 | 40110173 | ||
FJ210936.1 | - | 3 | 316 |