Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272064.1 | NP_001258993.1 | 19 | 3340 | ||
NM_001272076.1 | NP_001259005.1 | 19 | 3022 | |||
NM_145076.4 | NP_659542.3 | 19 | 3442 | |||
GenBank mRNA | AK133144.1 | - | 19 | 3022 | ||
AK166538.1 | - | 15 | 2399 | |||
BC025482.1 | - | 10 | 1537 | |||
BC056959.1 | - | 19 | 3420 | 5698079 | ||
S78219.1 | - | 19 | 3340 | |||
S78221.1 | - | 19 | 3442 |