Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009399.3 | NP_033425.3 | 10 | 1628 | ||
GenBank mRNA | AF019046.1 | - | 10 | 1623 | ||
AK138990.1 | - | 10 | 1633 | |||
BC019185.1 | - | 10 | 1625 | 5025470 |