Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011631.1 | NP_035761.1 | 16 | 2317 | ||
GenBank mRNA | AK005469.1 | - | 3 | 245 | ||
AK146885.1 | - | 16 | 2383 | |||
AK150811.1 | - | 16 | 2337 | |||
AK151413.1 | - | 16 | 2334 | |||
AK153550.1 | - | 16 | 2337 | |||
AK159974.1 | - | 16 | 2342 | |||
AK160093.1 | - | 16 | 2336 | |||
AK160827.1 | - | 1757 | ||||
AK165975.1 | - | 16 | 2337 | |||
AK168952.1 | - | 16 | 2337 | |||
AK169090.1 | - | 16 | 2337 | |||
AK209518.1 | - | 1 | 131 | |||
AK216243.1 | - | 1 | 131 | |||
BC010445.1 | - | 16 | 2249 | 3587178 | ||
BC011439.1 | - | 16 | 2323 | 3707817 | ||
J03297.1 | - | 16 | 2317 |