Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122754.2 | NP_001116226.1 | 5 | 512 | ||
NM_001289495.1 | NP_001276424.1 | 5 | 529 | |||
NM_013698.3 | NP_038726.2 | 4 | 596 | |||
GenBank mRNA | AK138069.1 | - | 5 | 509 | ||
AK138786.1 | - | 5 | 526 | |||
BC120552.1 | - | 4 | 499 | 40129475 | ||
BC137710.1 | - | 4 | 501 | 8860729 | ||
D43963.1 | - | 4 | 596 | |||
D83201.1 | - | 4 | 536 | |||
D83202.1 | - | 4 | 699 | |||
U16145.1 | - | 5 | 477 | |||
U19607.1 | - | 5 | 512 |