Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282014.1 | NP_001268943.1 | 8 | 1817 | ||
NM_001282015.1 | NP_001268944.1 | 8 | 1811 | |||
NM_031202.3 | NP_112479.1 | 8 | 1832 | |||
GenBank mRNA | AK148332.1 | - | 8 | 1817 | ||
AK148370.1 | - | 8 | 1723 | |||
AK148441.1 | - | 8 | 1723 | |||
AK177043.1 | - | 3 | 215 | |||
AK178803.1 | - | 3 | 215 | |||
AK179534.1 | - | 302 | ||||
AK188091.1 | - | 3 | 318 | |||
AK189488.1 | - | 3 | 215 | |||
AK190379.1 | - | 2 | 298 | |||
AK192864.1 | - | 3 | 321 | |||
AK192902.1 | - | 2 | 293 | |||
AK193585.1 | - | 3 | 318 | |||
AK194794.1 | - | 2 | 295 | |||
AK194981.1 | - | 2 | 298 | |||
AK196132.1 | - | 2 | 295 | |||
AK197740.1 | - | 3 | 318 | |||
AK198402.1 | - | 2 | 309 | |||
AK198835.1 | - | 3 | 318 | |||
AK198900.1 | - | 2 | 300 | |||
AK199149.1 | - | 3 | 318 | |||
AK199163.1 | - | 3 | 318 | |||
AK199296.1 | - | 3 | 311 | |||
AK199330.1 | - | 2 | 294 | |||
AK199776.1 | - | 3 | 318 | |||
AK199872.1 | - | 3 | 318 | |||
AK200013.1 | - | 3 | 318 | |||
AK200321.1 | - | 3 | 302 | |||
AK200365.1 | - | 3 | 318 | |||
AK200395.1 | - | 3 | 318 | |||
AK201323.1 | - | 2 | 285 | |||
AK202380.1 | - | 3 | 312 | |||
AK203562.1 | - | 2 | 273 | |||
AK204301.1 | - | 3 | 311 | |||
AK204415.1 | - | 2 | 287 | |||
AK207921.1 | - | 3 | 331 | |||
BC076598.1 | - | 8 | 1815 | 30613975 | ||
X03687.1 | - | 8 | 1717 |