Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083319.1 | NP_001076788.1 | 15 | 1832 | ||
NM_013699.2 | NP_038727.1 | 14 | 1724 | |||
GenBank mRNA | AB221581.1 | - | 13 | 1312 | ||
AK079815.1 | - | 7 | 559 | |||
AK140942.1 | - | 9 | 729 | |||
AK146040.1 | - | 3 | 344 | |||
AY182062.1 | - | 14 | 1421 | |||
BC034749.1 | - | 13 | 2051 | |||
BC139828.1 | - | 13 | 1341 | 40142981 | ||
BC141082.1 | - | 13 | 1403 | 9055694 | ||
U20086.1 | - | 13 | 1315 |