Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011674.4 | NP_035804.2 | 4 | 1083 | ||
GenBank mRNA | AK128994.1 | - | 4 | 1079 | ||
AK137364.1 | - | 4 | 1084 | |||
AK144005.1 | - | 4 | 1083 | |||
AK163650.1 | - | 4 | 1045 | |||
BC016885.1 | - | 1078 | 4223057 | |||
X92122.1 | - | 3 | 1408 |