Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011678.2 | NP_035808.2 | 10, 11 | 1375 | ||
GenBank mRNA | AK143582.1 | - | 10, 11 | 1310 | ||
AK149964.1 | - | 10, 11 | 1333 | |||
AK164245.1 | - | 1, 2 | 150 | |||
AK169372.1 | - | 10, 11 | 1354 | |||
AK169933.1 | - | 10, 11 | 1345 | |||
AK171271.1 | - | 10, 11 | 1345 | |||
AK171633.1 | - | 10, 11 | 1328 | |||
AK220512.1 | - | 10, 11 | 1332 | |||
BC011341.1 | - | 9, 10 | 1152 | 3661165 | ||
BC066180.1 | - | 10, 11 | 1285 | |||
BC066865.1 | - | 10, 11 | 1285 | |||
L00681.1 | - | 10, 11 | 1338 |