Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023478.2 | NP_075967.2 | 5 | 705 | ||
GenBank mRNA | AF222750.2 | - | 5 | 626 | ||
AK003737.1 | - | 3 | 216 | |||
BC125336.1 | - | 5 | 705 | 40129851 | ||
BC125338.1 | - | 5 | 705 | 40129853 | ||
BY703997.1 | - | 3 | 216 | |||
DQ387454.1 | - | 5 | 655 |