Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009483.1 | NP_033509.1 | 5 | 758 | ||
GenBank mRNA | AK080370.1 | - | 5 | 777 | ||
AK144654.1 | - | 5 | 784 | |||
AK164941.1 | - | 5 | 790 | |||
BC053433.1 | - | 5 | 758 | 6516368 |