Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145471.2 | NP_663446.1 | 6 | 1570 | ||
GenBank mRNA | AK036278.1 | - | 4 | 1443 | ||
AK047414.1 | - | 4 | 1115 | |||
AK053747.1 | - | 1680 | ||||
AK132158.1 | - | 4 | 2134 | |||
AK149655.1 | - | 4 | 1533 | |||
AK159836.1 | - | 4 | 1513 | |||
AK172871.1 | - | 4 | 2109 | |||
AY239225.1 | - | 4 | 1804 | |||
AY239226.1 | - | 6 | 1486 | |||
AY344668.1 | - | 4 | 1120 | |||
BC022126.1 | - | 6 | 1603 | 5097371 | ||
BC033482.1 | - | 3 | 1537 | |||
BC138217.1 | - | 4 | 1744 | 8861236 | ||
BC145203.1 | - | 4 | 1181 |