Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011714.2 | NP_035844.2 | 12 | 3502 | ||
GenBank mRNA | AF084480.1 | - | 12 | 3461 | ||
AK037737.1 | - | 6 | 710 | |||
BC141399.1 | - | 12 | 3405 | 9056012 | ||
BC171975.1 | - | 12 | 3404 |