Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153416.2 | NP_700465.2 | 1 | 184 | ||
GenBank mRNA | AK034591.1 | - | 1 | 79 | ||
AK079849.1 | - | 1 | 121 | |||
AK083401.1 | - | 1 | 97 | |||
AK083537.1 | - | 1 | 105 | |||
AK087134.1 | - | 1 | 159 | |||
AK088247.1 | - | 1 | 101 | |||
AK134406.1 | - | 1 | 75 | |||
AK165243.1 | - | 1 | 99 | |||
AK166118.1 | - | 1 | 99 | |||
BC018191.1 | - | 1 | 85 | 3994248 | ||
BC025501.1 | - | 1 | 131 | 5321013 |