Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flywch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153791.2 | NP_722486.2 | 3, 4 | 957 | ||
GenBank mRNA | AK034869.1 | - | 2, 3 | 551 | ||
AK052314.1 | - | 1, 2 | 169 | |||
AK129391.1 | - | 1, 2 | 115 | |||
AK168389.1 | - | 3, 4 | 892 | |||
BC038031.1 | - | 2, 3 | 1249 | 4021754 | ||
CA321280.1 | - | 2, 3 | 618 |