Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027060.1 | NP_081336.1 | 10 | 1767 | ||
NM_172618.2 | NP_766206.1 | 10 | 1767 | |||
GenBank mRNA | AK006275.1 | - | 5 | 653 | ||
AK029578.1 | - | 10 | 1930 | |||
AK029907.1 | - | 3 | 407 | |||
AK052664.1 | - | 10 | 1651 | |||
AK153973.1 | - | 10 | 1694 | |||
BC037031.1 | - | 8 | 1234 | |||
BC057897.1 | - | 10 | 1645 | 5031346 |