Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027060.1 | NP_081336.1 | 6 | 1233 | ||
NM_172618.2 | NP_766206.1 | 6 | 1233 | |||
GenBank mRNA | AK029578.1 | - | 6 | 1396 | ||
AK052664.1 | - | 6 | 1117 | |||
AK087881.1 | - | 6 | 1135 | |||
AK153973.1 | - | 6 | 1160 | |||
AK173285.1 | - | 3 | 441 | |||
AK192309.1 | - | 1 | 32 | |||
BC037031.1 | - | 4 | 700 | |||
BC057897.1 | - | 6 | 1111 | 5031346 |