Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172619.3 | NP_766207.2 | 3 | 380 | ||
GenBank mRNA | AK034565.1 | - | 3 | 355 | ||
AK034843.1 | - | 3 | 360 | |||
AK042525.1 | - | 3 | 368 | |||
AK046333.1 | - | 3 | 368 | |||
AK076334.1 | - | 3 | 354 | |||
BC056427.1 | - | 3 | 350 | |||
BC082773.1 | - | 3 | 372 | |||
BC100558.1 | - | 2 | 332 | |||
BC145353.1 | - | 2 | 287 | |||
BC145354.1 | - | 2 | 287 | |||
BC145355.1 | - | 2 | 287 |