Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013854.1 | NP_038882.1 | 17 | 1926 | ||
GenBank mRNA | AK145549.1 | - | 8 | 1078 | ||
AK153057.1 | - | 2 | 142 | |||
AK183892.1 | - | 1 | 27 | |||
BC040783.1 | - | 6 | 778 | |||
BC046965.1 | - | 17 | 1897 | 6485365 | ||
BC063094.1 | - | 17 | 1926 | 6516514 |