Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028639.3 | NP_082915.1 | 19 | 2648 | ||
GenBank mRNA | AK040578.1 | - | 19 | 2651 | ||
AK040610.1 | - | 19 | 2619 | |||
AK050263.1 | - | 19 | 2635 | |||
AK080284.1 | - | 19 | 2627 | |||
AK153704.1 | - | 19 | 2630 | |||
AK165244.1 | - | 12 | 1310 | |||
BC023773.1 | - | 15 | 1606 | |||
BC025613.1 | - | 1 | 131 | |||
BC042512.1 | - | 19 | 2325 | 4223801 | ||
BC049254.1 | - | 3 | 371 | |||
BC057906.1 | - | 2 | 271 |